Merck
  • Evolutionary Gain of Dbx1 Expression Drives Subplate Identity in the Cerebral Cortex.
Evolutionary Gain of Dbx1 Expression Drives Subplate Identity in the Cerebral Cortex.
Cell reports (2019-10-17)
Yoko Arai, Andrzej W Cwetsch, Eva Coppola, Sara Cipriani, Hidenori Nishihara, Hiroaki Kanki, Yoann Saillour, Betty Freret-Hodara, Annie Dutriaux, Norihiro Okada, Hideyuki Okano, Colette Dehay, Jeannette Nardelli, Pierre Gressens, Tomomi Shimogori, Giuseppe D'Onofrio, Alessandra Pierani
ABSTRACT
Changes in transcriptional regulation through cis-regulatory elements are thought to drive brain evolution. However, how this impacts the identity of primate cortical neurons is still unresolved. Here, we show that primate-specific cis-regulatory sequences upstream of the Dbx1 gene promote human-like expression in the mouse embryonic cerebral cortex, and this imparts cell identity. Indeed, while Dbx1 is expressed in highly restricted cortical progenitors in the mouse ventral pallium, it is maintained in neurons in primates. Phenocopy of the primate-like Dbx1 expression in mouse cortical progenitors induces ectopic Cajal-Retzius and subplate (SP) neurons, which are transient populations playing crucial roles in cortical development. A conditional expression solely in neurons uncouples mitotic and postmitotic activities of Dbx1 and exclusively promotes a SP-like fate. Our results highlight how transcriptional changes of a single fate determinant in postmitotic cells may contribute to the expansion of neuronal diversity during cortical evolution.
Sigma-Aldrich
Anti-Reelin Antibody, a.a. 164-496 mreelin, clone G10, clone G10, Chemicon®, from mouse
Sigma-Aldrich
Anti-Neural Cell Adhesion Molecule L1 Antibody, clone 324, clone 324, Chemicon®, from rat
Sigma-Aldrich
Anti-GABA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Reelin Antibody, a.a. 164-189 mreelin, clone 142, clone 142, Chemicon®, from mouse