Merck
  • Odorants from the Thermal Treatment of Hydrolyzed Mushroom Protein and Cysteine Enhance Saltiness Perception.
Odorants from the Thermal Treatment of Hydrolyzed Mushroom Protein and Cysteine Enhance Saltiness Perception.
Journal of agricultural and food chemistry (2019-10-09)
Jordan Lopez, Trent Kerley, Lindsay Jenkinson, Curtis R Luckett, John P Munafo
ABSTRACT
Innovative approaches to develop flavors with high sensory appeal are critical in encouraging increased consumer preference and adoption of low sodium foods. Gas chromatography-olfactometry, coupled with stable isotope dilution assays and sensory experiments, led to the identification of the odorants responsible for an enhancement in saltiness perception of chicken broth prepared with thermally treated enzymatically hydrolyzed mushroom protein and cysteine, then reacted under kitchen-like cooking conditions. Comparative aroma extract dilution analysis revealed 36 odorants with flavor dilution factors between a range of 1 and 256. Sixteen odorants were quantitated and odor activity values (OAVs) calculated. Important odorants included 2-furfurylthiol (coffee, OAV 610), 1-(2-furyl)ethanethiol (meaty, OAV 78), 3-sulfanylpentan-2-one (catty, OAV 42), sotolon (maple, OAV 20), indole (animal, OAV 8), 2-methyl-3-(methyldithio)furan (meaty, OAV 3), and p-cresol (barnyard, OAV 1). An odor simulation model was evaluated in two consumer sensory studies. These studies confirmed that the addition of the aroma model increased the perceived saltiness of low sodium chicken broth (p < 0.05).
Sigma-Aldrich
Lawesson reagent, 97%
Sigma-Aldrich
Pentadecane-d32, 98 atom % D, 98% (CP)
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
(±)-1-(2-Furyl)ethanol, ≥99.0% (GC)
Sigma-Aldrich
Maltose solution, BioReagent, for molecular biology, ~20% in H2O