Merck
  • H3K4 dimethylation at FosB promoter in the striatum of chronic stressed rats promotes morphine-induced conditioned place preference.
H3K4 dimethylation at FosB promoter in the striatum of chronic stressed rats promotes morphine-induced conditioned place preference.
PloS one (2019-08-24)
Minghui Chen, Xiaojie Zhang, Wei Hao
ABSTRACT
Expression of FosB gene in striatum is essential in addiction establishment. Activated glucocorticoid receptors (GRs) induce FosB gene expression in response to stressor. Therefore, elevation of FosB expression in striatum serves as one mechanism by which stress increases risk for addiction. In this study, adult male Sprague-Dawley rats were used to investigate whether chronic stress result in histone modifications at FosB gene promoter in striatum and how these histone modifications affect FosB expression and the establishment of addiction behavior after administration of drugs of abuse. Animals were randomly assigned to three groups: Electric foot shock (EFS) group received 7-day EFS to induce chronic stress; electric foot shock plus mifepristone (EFS + Mif) group were injected with mifepristone, a nonspecific GRs antagonist, before EFS; control group did not receive any EFS. All groups then received 2-day conditioned place preference (CPP) training with morphine (5 mg/kg body weight) to test vulnerability to drug addiction. Before and after morphine administration, FosB mRNA in striatum was quantified by real-time RT-PCR. Levels of histone H3/H4 acetylation and histone H3K4 dimethylation at FosB promoter in striatum after morphine administration were measured by using chromatin immunoprecipitation (ChIP) plus real-time PCR. EFS group had stronger place preference to morphine and had significantly higher level of FosB mRNA in striatum than the other two groups. H3K4 dimethylation was 2.6-fold higher in EFS group than control group, while no statistical difference in H3/H4 acetylation. Mifepristone administration before EFS decreased histone H3K4 dimethylation and FosB mRNA in striatum, and also diminished morphine-induced conditioned place preference. Altogether, increased level of H3K4 dimethylation at FosB promoter in striatum is partially dependent on the activation of GR and responsible for the elevated level of morphine-induced FosB mRNA in chronic stressed animals.
Sigma-Aldrich
LiCl Immune Complex Wash Buffer, For use in Chromatin Immunoprecipitation assays.
Sigma-Aldrich
Lys-Lys-Lys-Lys, ≥95% (TLC)