Merck
  • Niemann-Pick Type C Disease Reveals a Link between Lysosomal Cholesterol and PtdIns(4,5)P2 That Regulates Neuronal Excitability.
Niemann-Pick Type C Disease Reveals a Link between Lysosomal Cholesterol and PtdIns(4,5)P2 That Regulates Neuronal Excitability.
Cell reports (2019-05-30)
Oscar Vivas, Scott A Tiscione, Rose E Dixon, Daniel S Ory, Eamonn J Dickson
ABSTRACT
There is increasing evidence that the lysosome is involved in the pathogenesis of a variety of neurodegenerative disorders. Thus, mechanisms that link lysosome dysfunction to the disruption of neuronal homeostasis offer opportunities to understand the molecular underpinnings of neurodegeneration and potentially identify specific therapeutic targets. Here, using a monogenic neurodegenerative disorder, NPC1 disease, we demonstrate that reduced cholesterol efflux from lysosomes aberrantly modifies neuronal firing patterns. The molecular mechanism linking alterations in lysosomal cholesterol egress to intrinsic tuning of neuronal excitability is a transcriptionally mediated upregulation of the ABCA1 transporter, whose PtdIns(4,5)P2-floppase activity decreases plasma membrane PtdIns(4,5)P2. The consequence of reduced PtdIns(4,5)P2 is a parallel decrease in a key regulator of neuronal excitability, the voltage-gated KCNQ2/3 potassium channel, which leads to hyperexcitability in NPC1 disease neurons. Thus, cholesterol efflux from lysosomes regulates PtdIns(4,5)P2 to shape the electrical and functional identity of the plasma membrane of neurons in health and disease.
Sigma-Aldrich
Filipin complex from Streptomyces filipinensis, ≥70% (UV)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Anti-SYNJ1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Avanti
17:0-20:4 PI(4)P, Avanti Research - A Croda Brand LM1901, powder
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, non-essential amino acids and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt ≥300,000, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Avanti
17:0-20:4 PI(4,5)P2, Avanti Research - A Croda Brand LM1904, powder