- CCAAT/enhancer-binding protein mRNA is translated into multiple proteins with different transcription activation potentials.
CCAAT/enhancer-binding protein mRNA is translated into multiple proteins with different transcription activation potentials.
The CCAAT/enhancer-binding protein (C/EBP) alpha is a leucine zipper protein that is preferentially expressed in certain cell types, such as adipocytes and hepatocytes. Here we show that C/EBP alpha mRNA is translated into two major proteins, C/EBP-42 and C/EBP-30, that differ in their content of N-terminal amino acid sequences. These results are best explained by a ribosome-scanning mechanism in which a fraction of ribosomes ignore the first two AUGs and initiate translation at an AUG located 351 nt downstream of the first one. Because C/EBP-30, the translation product initiated at the third AUG, is devoid of the potent transcription-activation domain contained in C/EBP-42, the former protein stimulates transcription from the mouse albumin promoter much less efficiently than the latter. The gene encoding the liver-enriched transcriptional-activator protein LAP (C/EBP-beta) has also been shown to issue two proteins, LAP and the liver-enriched transcriptional-inhibitory protein LIP, with different transcription-activation potentials. The production of multiple proteins from a single mRNA is not only shared between different C/EBP family members but also appears to be conserved in vertebrate evolution.