Merck
Pathogenic Escherichia coli.
Nature reviews. Microbiology (2004-03-26)
James B Kaper, James P Nataro, Harry L Mobley
ABSTRACT
Few microorganisms are as versatile as Escherichia coli. An important member of the normal intestinal microflora of humans and other mammals, E. coli has also been widely exploited as a cloning host in recombinant DNA technology. But E. coli is more than just a laboratory workhorse or harmless intestinal inhabitant; it can also be a highly versatile, and frequently deadly, pathogen. Several different E. coli strains cause diverse intestinal and extraintestinal diseases by means of virulence factors that affect a wide range of cellular processes.
Sigma-Aldrich
Escherichia coli FISH probe -ATTO488, Probe for fluorescence in situ hybridization (FISH), 20µM in water
Sigma-Aldrich
Microbial DNA standard from Escherichia coli, Suitable for PCR, sequencing and NGS, 10 ng/μL