Merck
  • Measurement of lidocaine and 2,6-dimethylaniline in minipig plasma, skin, and dermal tapes using UHPLC with electrospray MS/MS.
Measurement of lidocaine and 2,6-dimethylaniline in minipig plasma, skin, and dermal tapes using UHPLC with electrospray MS/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2018-05-11)
Qian Li, Tobias Magers, Brad King, Brian J Engel, Ray Bakhtiar, Charisse Green, Ronald Shoup
ABSTRACT
Sensitive LC-MS/MS methods were developed to measure lidocaine and its metabolite 2,6-dimethylaniline (2,6-DMA) with application to transdermal studies. The methods for lidocaine in minipig plasma, tissue biopsies, and dermal tapes utilized mixed mode/SCX solid phase extraction, with lower quantitation limits of 25 pg/mL in plasma, 15 ng/g tissue, and 5 ng/tape. 2,6-DMA was measured in plasma and skin tissue homogenates by ultrafiltration and (for tissue) by further derivatization with 4-methoxybenzoyl chloride to form the corresponding benzamide derivative, which extended the lower limit of quantitation to 200 pg/mL. The methods allowed local measurement of lidocaine in stratum corneum, punch biopsies, and plasma and of 2,6-DMA in plasma and biopsies obtained from minipigs dosed with experimental transdermal formulations. Quantitation limits were approximately 7-fold lower than previously reported for lidocaine and 3-fold lower for 2,6-DMA.
Sigma-Aldrich
4-Methoxybenzoyl chloride, 99%
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride