Merck
  • Olig2 overexpression induces the in vitro differentiation of neural stem cells into mature oligodendrocytes.
Olig2 overexpression induces the in vitro differentiation of neural stem cells into mature oligodendrocytes.
Stem cells (Dayton, Ohio) (2005-10-29)
Sjef Copray, Veerakumar Balasubramaniyan, Josien Levenga, Jorick de Bruijn, Robert Liem, Erik Boddeke
ABSTRACT
Differentiation induction of neural stem cells (NSCs) into oligodendrocytes during embryogenesis is the result of a complex interaction between local induction factors and intracellular transcription factors. At the early stage of differentiation, in particular, the helix-loop-helix transcription factors Olig1 and Olig2 have been shown to be essential for oligodendrocyte lineage determination. In view of the possible application of NSCs as a source for remyelinating cell transplants in demyelinating diseases (e.g., multiple sclerosis), in vitro procedures need to be developed to drive the oligodendrocyte differentiation process. Mere culture in medium supplemented with major embryonic oligodendrogenic induction factors, such as Sonic hedgehog, results in oligodendrocyte differentiation of only about 10% of NSCs. We previously showed that induction of Olig1 expression by gene transfection could indeed initiate the first stage of oligodendrocyte differentiation in NSCs, but appeared to be unable to generate fully mature, functional oligodendrocytes. In this study, we transfected NSCs isolated from the embryonic mouse brain with the Olig2 gene and found that the introduced overexpression of Olig2 could induce the development of fully mature oligodendrocytes expressing the transcription factor Nkx2.2 and all major myelin-specific proteins. Moreover, Olig2-transfected NSCs, in contrast to nontransfected NSCs, developed into actively remyelinating oligodendrocytes after transplantation into the corpus callo-sum of long-term cuprizonefed mice, an animal model for demyelination. Our results show that transfection of genes encoding for oligodendrogenic transcription factors can be an efficient way to induce the differentiation of NSCs into functional oligodendrocytes.
Sigma-Aldrich
Anti-A2B5 Antibody, clone A2B5-105, clone A2B5-105, Chemicon®, from mouse
Sigma-Aldrich
Anti-Nestin Antibody, clone rat-401, clone rat-401, Chemicon®, from mouse
Sigma-Aldrich
Anti-Galactocerebroside Antibody, clone mGalC, clone mGalC, Chemicon®, from mouse
Sigma-Aldrich
Anti-Oligodendrocytes Antibody, clone CE-1, ascites fluid, clone CE-1, Chemicon®
Sigma-Aldrich
Monoclonal Anti-β-Tubulin III antibody produced in mouse, clone SDL.3D10, ascites fluid
Sigma-Aldrich
Anti-Olig1 Antibody, Chemicon®, from rabbit