
USGS32 (nitrogen and oxygen isotopes in nitrate)

Synonym(s):
Potassium nitrate
Empirical Formula (Hill Notation):
KNO3
CAS Number:
Molecular Weight:
101.10
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Quality Segment
Isotope
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing