
Tulobuterol hydrochloride

Synonym(s):
2-tert-Butylamino-1-(2-chlorophenyl)ethanol hydrochloride
Empirical Formula (Hill Notation):
C12H18ClNO · HCl
CAS Number:
Molecular Weight:
264.19
Products
Brand
Formula Weight
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing