Skip to Content
Merck
Trigalacturonic acid ≥90% (HPLC)

Trigalacturonic acid

Trigalacturonic acid ≥90% (HPLC)
Synonym(s):
α-D-GalA-(1→4)-α-D-GalA-(1→4)-α-D-GalA, α-D-GalA-(1→4)-α-D-GalA-(1→4)-D-GalA
Empirical Formula (Hill Notation):
C18H26O19
CAS Number:
Molecular Weight:
546.39

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing