Tioconazole
Synonym(s):
1-[2-[(2-Chloro-3-thienyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole, Thioconazole
Empirical Formula (Hill Notation):
C16H13Cl3N2OS
CAS Number:
Molecular Weight:
387.71
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing