
Tinidazole

Synonym(s):
1-[2-(Ethylsulfonyl)ethyl]-2-methyl-5-nitroimidazole
Empirical Formula (Hill Notation):
C8H13N3O4S
CAS Number:
Molecular Weight:
247.27
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3