
Stearic acid 50

Synonym(s):
Palmitic acid – stearic acid mixture, Fatty acids C16-18, Stearic acid - palmitic acid mixture
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
Feature
Physical Form
Agency
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing