
Soyasaponin I

Synonym(s):
SCM 3B, Soyasaponin Bb
Empirical Formula (Hill Notation):
C48H78O18
CAS Number:
Molecular Weight:
943.12
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Research Area
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing