Skip to Content
Merck
Sodium 1-heptanesulfonate monohydrate ≥99.0% (T)

Sodium 1-heptanesulfonate monohydrate

Sodium 1-heptanesulfonate monohydrate ≥99.0% (T)
Synonym(s):
1-Heptanesulfonic acid sodium salt monohydrate
Linear Formula:
CH3(CH2)5CH2SO3Na · H2O
CAS Number:
Molecular Weight:
220.26
EC Number:

Products

Product Category
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Technique
Showing 1-3 of 3