
Shatavarin IV

Synonym(s):
3β,5β,25S)-Spirostan-3-yl O-6-deoxy-α-L-mannopyranosyl-(1→4)-O-[β-D-glucopyranosyl-(1→2)]-β-D-glucopyranoside, Asparanin B, Curillin H
Empirical Formula (Hill Notation):
C45H74O17
CAS Number:
Molecular Weight:
887.06
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing