Skip to Content
Merck
(S)-Duloxetine hydrochloride ≥98% (HPLC)

(S)-Duloxetine hydrochloride

(S)-Duloxetine hydrochloride ≥98% (HPLC)
Synonym(s):
(γS)-N-Methyl-γ-(1-naphthalenyloxy)-2-thiophenepropanamine hydrochloride, (+)-(S)-N-Methyl-γ-(1-naphthyloxy)-2-thiophenepropylamine hydrochloride, (+)-N-Methyl-3-(1-naphthalenyloxy)-3-(2-thienyl)propanamine hydrochloride, LY-248686 hydrochloride
Empirical Formula (Hill Notation):
C18H19NOS · HCl
CAS Number:
Molecular Weight:
333.88

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing