Skip to Content
Merck
Scandium(III) triflate 99.995% trace metals basis

Scandium(III) triflate

Scandium(III) triflate 99.995% trace metals basis
Synonym(s):
Scandium(III) trifluoromethanesulfonate, Trifluoromethanesulfonic acid scandium(III) salt
Linear Formula:
Sc(SO3CF3)3
CAS Number:
Molecular Weight:
492.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing