Skip to Content
Merck
(R)-(−)-3-Hydroxybutyric acid sodium salt optical purity ee: 99% (GLC)

(R)-(−)-3-Hydroxybutyric acid sodium salt

(R)-(−)-3-Hydroxybutyric acid sodium salt optical purity ee: 99% (GLC)
Synonym(s):
Sodium (R)-3-hydroxybutyrate
Linear Formula:
CH3CH(OH)CH2CO2Na
CAS Number:
Molecular Weight:
126.09

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing