
Progesterone-Water Soluble

Synonym(s):
Progesterone: HBC complex, Progesterone: methyl-β-cyclodextrin complex
Products
Product Category
Brand
Biological Source
Gene Alias
Physical Form
Product Line
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing