
PPT

Synonym(s):
2,8-Bis(diphenylphosphoryl)dibenzo[b,d]thiophene
Empirical Formula (Hill Notation):
C36H26O2P2S
CAS Number:
Molecular Weight:
584.60
Products
Brand
Color
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing