![Potassium nitrate NIST® SRM® 193](/deepweb/assets/sigmaaldrich/product/structures/204/149/f70096c8-36e0-4faa-aa55-413e3060c0a8/640/f70096c8-36e0-4faa-aa55-413e3060c0a8.png)
Potassium nitrate
![Potassium nitrate NIST® SRM® 193](/deepweb/assets/sigmaaldrich/product/structures/204/149/f70096c8-36e0-4faa-aa55-413e3060c0a8/640/f70096c8-36e0-4faa-aa55-413e3060c0a8.png)
Synonym(s):
Potassium nitrate
Empirical Formula (Hill Notation):
KNO3
CAS Number:
Molecular Weight:
101.10
Products
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-14 of 14
Compare
Product Number
Description
Pricing