
Picosulfate for system suitability

Synonym(s):
Sodium picosulfate
Empirical Formula (Hill Notation):
C18H13NNa2O8S2
CAS Number:
Molecular Weight:
481.41
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing