Paroxetine hydrochloride hemihydrate
Synonym(s):
(3S-trans)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine hydrochloride hemihydrate
Empirical Formula (Hill Notation):
C19H20FNO3 · HCl · .5 H2O
CAS Number:
Molecular Weight:
374.83
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2