
N,N′-Bis(2,4-xylyl)formamidine

Synonym(s):
N,N′-Bis(2,4-dimethylphenyl)methanimidamide
Empirical Formula (Hill Notation):
C17H20N2
CAS Number:
Molecular Weight:
252.35
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing