
N-Methyl-5-(methylamino)-1H-imidazole-4-carboxamide

Synonym(s):
Theophyllidine
Empirical Formula (Hill Notation):
C6H10N4O
CAS Number:
Molecular Weight:
154.17
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Special Grade
Showing 1-1 of 1