
N-(Methoxysuccinyl)-Ala-Ala-Pro-Val-chloromethyl ketone

Synonym(s):
N-(Methoxysuccinyl)-L-alanyl-L-alanyl-L-prolyl-L-valine chloromethylketone
Empirical Formula (Hill Notation):
C22H35ClN4O7
CAS Number:
Molecular Weight:
502.99
Products
Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing