
N-Ethylisopropylamine

Synonym(s):
1-Methyldiethylamine
Linear Formula:
(CH3)2CHNHC2H5
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing