
Nandrolone laurate

Synonym(s):
19-Nortestosterone 17-dodecanoate, Laurabolin, Nandrolone laurate
Empirical Formula (Hill Notation):
C30H48O3
CAS Number:
Molecular Weight:
456.70
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing