
Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide

Synonym(s):
FDAA, Marfey’s reagent
Empirical Formula (Hill Notation):
C9H9FN4O5
CAS Number:
Molecular Weight:
272.19
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing