N1-(3-Trimethoxysilylpropyl)diethylenetriamine
Synonym(s):
2-[2-(3-Trimethoxysilylpropylamino)ethylamino]ethylamine, 3-[2-(2-Aminoethylamino)ethylamino]propyltrimethoxysilane
Linear Formula:
(CH3O)3Si(CH2)3NHCH2CH2NHCH2CH2NH2
CAS Number:
Molecular Weight:
265.43
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing