Skip to Content
Merck
Methyl acetoacetate sodium salt ≥97.0% (T)

Methyl acetoacetate sodium salt

Methyl acetoacetate sodium salt ≥97.0% (T)
Synonym(s):
Sodium methyl acetoacetate
Linear Formula:
CH3COCHNaCOOCH3
CAS Number:
Molecular Weight:
138.10
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing