![Maltotriose ≥90% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/109/771/3c6c3dbc-8dcd-4b85-a401-ae34c4c8ba66/640/3c6c3dbc-8dcd-4b85-a401-ae34c4c8ba66.png)
Maltotriose
![Maltotriose ≥90% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/109/771/3c6c3dbc-8dcd-4b85-a401-ae34c4c8ba66/640/3c6c3dbc-8dcd-4b85-a401-ae34c4c8ba66.png)
Synonym(s):
α-D-Glc-(1→4)-α-D-Glc-(1→4)-D-Glc, O-α-DD-Glucopyranosyl-(1→4)-O-α-D-glucopyranosyl-(1→4)-D-glucose, Maltotriose
Empirical Formula (Hill Notation):
C18H32O16
CAS Number:
Molecular Weight:
504.44
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Optical Isomer
Showing 1-2 of 2
Compare
Product Number
Description
Pricing