
Levodropropizine

Synonym(s):
Levodropropizine, (2S)-3-(4-Phenyl-1-piperazinyl)-1,2-propanediol
Empirical Formula (Hill Notation):
C13H20N2O2
CAS Number:
Molecular Weight:
236.31
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing