
Lauryl gallate

Synonym(s):
Dodecyl gallate, Lauryl gallate
Empirical Formula (Hill Notation):
C19H30O5
CAS Number:
Molecular Weight:
338.44
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing