
Koenigicine

Synonym(s):
3,11-Dihydro-8,9-dimethoxy-3,3,5-trimethylpyrano[3,2-a]carbazole, Kenimbidine, Koenidine, Koenimbidine, NSC 127151
Empirical Formula (Hill Notation):
C20H21NO3
CAS Number:
Molecular Weight:
323.39
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing