Skip to Content
Merck
Isobutyl acetate analytical standard

Isobutyl acetate

Isobutyl acetate analytical standard
Synonym(s):
Acetic acid isobutyl ester, Isobutyl acetate
Linear Formula:
CH3COOCH2CH(CH3)2
CAS Number:
Molecular Weight:
116.16
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique