
Iscotrizinol

Synonym(s):
4,4-[[6-[[4-[[(1,1-Dimethylethyl)amino]carbonyl]phenyl]amino]-1,3,5-triazine-2,4-diyl]diimino]bisbenzoic acid 1,1′-bis(2-ethylhexyl) ester, 4,6-Bis[4-(2-ethylhexyloxycarbonyl)anilino]-2-[4-(tert-butylaminocarbonyl)anilino]-1,3,5-triazine, Bis(2-ethylhexyl) 4,4‘-[(6-{[4-(tert-butylcarbamoyl)phenyl]amino}-1,3,5-triazine-2,4-diyl)diimino]dibenzoate, Diethylhexyl butamido triazone
Empirical Formula (Hill Notation):
C44H59N7O5
CAS Number:
Molecular Weight:
765.98
Products
Brand
Formula Weight
Manufacturer
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing