
IMDM

Synonym(s):
IMDM, Iscove’s Modified Dulbecco’s Medium
Products
Product Category
Brand
Physical Form
Glucose (g/L)
L-Glutamine
Sodium Bicarbonate
Phenol Red
Stable Glutamine
Quality Segment
Sterilization Method
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing