
Ibandronate Related Compound A

Synonym(s):
3-[Methyl(pentyl)amino]propanoic acid hydrochloride, N-Methyl-N-pentyl-β-alanine hydrochloride, Ibanic acid hydrochloride
Empirical Formula (Hill Notation):
C9H19NO2 · HCl
CAS Number:
Molecular Weight:
209.71
Products
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2