
Hydrocortisone valerate

Synonym(s):
Hydrocortisone 17-valerate, 11β,17,21-Trihydroxypregn-4-ene-3,20-dione 17-valerate, Cortisol 17-valerate
Empirical Formula (Hill Notation):
C26H38O6
CAS Number:
Molecular Weight:
446.58
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing