
Furazolidone

Synonym(s):
3-(5-Nitrofurfurylideneamino)-2-oxazolidinone, Furazolidone
Empirical Formula (Hill Notation):
C8H7N3O5
CAS Number:
Molecular Weight:
225.16
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3