
Foscarnet impurity D

Synonym(s):
Triethyl phosphonoformate, Ethyl diethoxyphospinylformate
Linear Formula:
(C2H5O)2P(O)COOC2H5
CAS Number:
Molecular Weight:
210.16
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing