Foscarnet impurity B
Synonym(s):
1-Ethoxy-1-hydroxyphosphinecarboxylic acid 1-oxide disodium salt, Disodium (ethoxyoxydophosphanyl)formate
Empirical Formula (Hill Notation):
C3H5Na2O5P
CAS Number:
Molecular Weight:
198.02
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing