
Flurazepam HCl

Synonym(s):
Flurazepam dihydrochloride, 7-Chloro-1-(diethylamino)ethyl-5-(2-fluorophenyl)-3H-1,4-benzodiazepin-2(1H)-one dihydrochloride
Empirical Formula (Hill Notation):
C21H23ClFN3O · 2HCl
CAS Number:
Molecular Weight:
460.80
Products
Brand
Formula Weight
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing