
Flunitrazepam

Synonym(s):
5-(2-Fluorophenyl)-1-methyl-7-nitro-3H-1,4-benzodiazepin-2(1H)-one, Ro 5-4200
Empirical Formula (Hill Notation):
C16H12FN3O3
CAS Number:
Molecular Weight:
313.28
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing