Ethylene glycol-bis(β-aminoethyl ether)-N,N,N′,N′-tetraacetic acid tetrasodium salt
Synonym(s):
EGTA tetrasodium, Egtazic acid tetrasodium, Ethylene-bis(oxyethylenenitrilo)tetraacetic acid tetrasodium
Empirical Formula (Hill Notation):
C14H20N2O10Na4
CAS Number:
Molecular Weight:
468.28
Products
Brand
Formula Weight
Purity
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing