
Ethylene glycol

Synonym(s):
1,2-Ethanediol, Ethylene glycol
Linear Formula:
HOCH2CH2OH
CAS Number:
Molecular Weight:
62.07
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-11 of 11
Compare
Product Number
Description
Pricing