
Ethylenediaminetetraacetic acid dipotassium salt dihydrate

Synonym(s):
Ethylenediaminetetraacetic acid dipotassium salt dihydrate, (Ethylenedinitrilo)tetraacetic acid dipotassium salt, Dipotassium ethylenediaminetetraacetate dihydrate, EDTA dipotassium salt, Edathamil, Potassium ethylenediaminetetraacetate dibasic
Linear Formula:
HOOCCH2(KOOCCH2)NCH2CH2N(CH2COOK)CH2COOH · 2H2O
Products
Product Category
Brand
Biological Source
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing